Gebogen pincetten
Leverancier: HAMMACHER KARL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HWC071-10 HWC073-11 HWC075-13
232-2194EA 69.6 EUR
232-2194 232-0066 232-2154
Gebogen pincetten
Pincetten
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
  • Delicate
  • Resistant to corrosion
  • Anti-magnetic