Kühne gebogen pincetten
Leverancier: HAMMACHER KARL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HWC147-10 HWC149-16
232-2200EA 59.1 EUR
232-2200 232-2183
Kühne gebogen pincetten
Pincetten
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
  • For microscopic slides
  • Resistant to corrosion
  • Anti-magnetic
Order Now
Specificaties
Environmentally Preferable