Rechte pincetten met muizentanden
Leverancier: HAMMACHER KARL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HWC080-12 HWC081-13 HWC082-14
232-0067EA 83.7 EUR
232-0067 232-0068 232-2190
Rechte pincetten met muizentanden
Pincetten
WIRONIT steel (18/12 stainless steel with 18% chrome and 10 - 12 % nickel)
  • Teethed, straight
  • Resistant to corrosion
  • Anti-magnetic