Poedertrechters
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70894 71194
221-0160EA 3.2 EUR
221-0160 221-0163
Poedertrechters
Trechters Poedertrechters
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Used for transferring powders. Also suitable for liquids.