Funnels with short stem
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2140/10M 2140/11M 2140/12M 2140/14M 2140/01M 2140/06M 2140/02M 2140/03M
221-0006EA 25.8 EUR
221-0006 221-0020 221-0007 221-0008 221-0003 221-0005 221-0004 221-0017
Funnels with short stem
Trechters Filtertrechters Filtertrechters
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls.