Microtrechters
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F146850024 F146850028 F146850035
221-1248EA 80.1 EUR
221-1248 221-1249 221-1250
Microtrechters
Trechters Filtertrechters Filtertrechters
PP
  • Good chemical resistance
  • Autoclavable at 121 °C
Ideal for filling burettes and small bottles.  