Büchner trechters, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SF3A33 SF3A32 SF3B33
532-0097EA 121 EUR
532-0097 201-8681 201-8682
Büchner trechters, Quickfit®
Trechters Filtertrechters Büchner-trechters
PYREX®, borosilicaatglas.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc and integral ground glass cone.