Filtratietrechter Microcheck® II
Leverancier: Cytiva (Formerly Pall Lab)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4761 4763 4764 4761ME
516-0201EA 204 EUR
516-0201 516-0203 516-0204 516-1227
Filtratietrechter Microcheck® II
Trechters Membraanfiltertrechters
MicroCheck™ II beverage monitor, is an easy-to-use disposable filter funnel designed to meet the needs for microbial analysis of beverages.
  • Membrane diameter: 47 mm
  • Diameter: 6,1 cm
  • Membrane size: 0,45 µm
Polypropylene construction eliminates compatibility or extractables issues. Ergonomically designed units allow for removal of funnel cylinder with a squeeze. The base and cover convert to a petri dish for culturing the membrane in place, or the membrane can be cultured on a separate petri dish. Unit fits directly onto a standard manifold, or aMore Product Information