Cleanroom handschoenen, latex, steriel, Kimtech™ G3
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC1370S HC1390S
112-0491EA 0 EUR
112-0491 112-4830
Cleanroom handschoenen, latex, steriel, Kimtech™ G3
Handschoenen
These hand-specific gloves with beaded cuffs are specially washed to control particle levels.
  • Sterile, powder-free
  • Low protein and low endotoxin levels
  • Textured, non-slip surface provides exceptional wet/dry grip without compromising tactile sensitivity
  • Tapered, beaded cuff reduces roll down, makes donning easier, enhances cuff strength, and forms a better seal on garments
  • Sterilised by gamma irradiation
Suitable for use in Class 1, ISO 3, M1.5 environments.
Length: 300 mm
Certificaten: PPE Cat III (EU) 2016/425; EN 374-1 Type C (K), EN 374-5 (VIRUS); AQL 1.5
Verpakking: Elk paar is dubbel verpakt in een envelop in een zakje, 20 zakjes per buitenzak, 10 buitenzakken per doos (200 paar).