Latex handschoenen, Semperclean MC
Leverancier: SEMPERIT TECHN PROD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
814720622 814720722 814720702 814720802
112-5700EA 61.5 EUR
112-5700 112-5701 112-3130 112-3131
Latex handschoenen, Semperclean MC
Handschoenen
These hand-specific disposable gloves with beaded cuffs feature a special inner coating. Ideal for use in laboratories, electronic and pharmaceutical industries.
  • Non sterile, powder-free, micro-roughened finish
  • Extremely comfortable to wear and particularly easy to put on due to the special inner coating
  • Anatomical shape reduces hand fatigue
  • Good grip due to special surface treatment
  • Skin friendly due to cleaning and washing procedure
Ideal for use in laboratories, electronic and pharmaceutical industries.
Length: 270 - 285 mm
Thickness: 0,21 mm ± 0,05 mm
Colour: Natural
EN 455, EN 374, Cat. III; AQL 1.5
Certificaten: EN 455, EN 374; Cat. III; AQL 1,5