Onderzoekshandschoenen, nitril, Micro-Touch® Nitra-Tex™ en Nitra-Tex™ EP™
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Micro-Touch®
112-1850EA 20.1 EUR
112-1850 112-1848 112-1849 112-1846 112-1847 112-1844 112-1845 112-1853 112-1854 112-1843
Onderzoekshandschoenen, nitril, Micro-Touch® Nitra-Tex™ en Nitra-Tex™ EP™
Handschoenen
NitraTex® gloves are ideal for use in daily examinations and laboratory work in doctors' surgeries, laboratories and hospitals. At 300 mm, Micro-Touch® Nitra-Tex™ EP™ offers additional length for extra protection.
Certificaten: EN 455 (1-4), EN 374-1 Type B (KPT), EN 374-5, EN 421, Cat. III
Goedgekeurd voor voedselcontact