Chemisch bestendige handschoenen, AlphaTec® Solvex® 37-655 / 37-185
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
37-655/9 37-185/10 37-185/9 37-185/11 37-185/8 37-185/7
112-0283EA 0 EUR
112-0283 112-2321 112-1168 112-2927 112-2929 112-2928
Chemisch bestendige handschoenen, AlphaTec® Solvex® 37-655 / 37-185
Handschoenen
De Solvex®-handschoen is voorzien van een robuuste, chemicaliënbestendige nitrilcoating.
  • Fully reusable, with an unequalled abrasion resistance and mechanical protection
  • Flocked lining provides high comfort
  • Unlined version has no inner cotton flocking, reducing risk of lint contamination, and making it ideal for production areas sensitive to the introduction of external impurities
  • Sand-patch finish provides a smooth surface: Suitable for handling fragile items
These gloves are the ideal choice for safe handling in a wide range of work environments where harsh chemicals are present.
Thickness: 0,38 or 0,56 mm
Colour: Green
EN 374, EN 388; EN 1149, Cat. III, AQL 0.65
Certificaten: 37-185: EN ISO 374-1:2016 Type A (AGJKLPT), EN ISO 374-5:2016, EN 388:2016 (4102X), EN 1149; Cat. III
37-655: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN 388:2016 (4101X), EN 1149; Cat. III
Food contact approved.