Chemisch bestendige handschoenen, AlphaTec® Solvex® 37-900
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
37-900/10 37-900/11 37-900/7 37-900/8 37-900/9
112-0968EA 10.6 EUR
112-0968 112-0969 112-0970 112-0971 112-0972
Chemisch bestendige handschoenen, AlphaTec® Solvex® 37-900
Handschoenen
These cotton flock lined gloves feature a special nitrile coating and a textured finish ensuring a safe grip.
  • Density tested
  • Anatomical design, reduces hand fatigue
  • Sanitised/Actifresh: Contains fungicidal and bacteriostatic agent to maintain hand hygiene and protect against skin irritation
Ideal for use in chemical processing, manufacture of semiconductors and agriculture (handling of pesticides).
Thickness: 0,425 mm
Colour: Red
EN 388 (4102), EN 374 (AKL), Cat. III; AQL 0.065
Certificaten: EN ISO 374-1:2016 Type A (AKLJOPT), EN ISO 374-5:2016 VIRUS, EN 388:2016 (4101X), Cat. III.