Chemisch bestendige handschoenen, AlphaTec® 87-900
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87-900/6.5-7 87-900/7.5-8 87-900/8.5-9 87-900/7
112-0305EA 0 EUR
112-0305 112-0306 112-0307 ANSE87-900/7
Chemisch bestendige handschoenen, AlphaTec® 87-900
Handschoenen
Heavyweight latex/neoprene gloves with pure cotton flock lining and embossed lozenge finish.
  • Reliable protection against a very wide range of chemicals
  • Highly resistant to salts, detergents and ketones
  • Excellent wet and dry grip
For chemical, electronic and aircraft industries.
Certificaten: EN 388:2016 (2120A), EN ISO 374-1 Type A (AKLNPT), EN ISO 374-5 VIRUS, EN 421; Cat. III