Snijbestendige handschoenen, HyFlex® 70-215 / 70-225
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70-215/7 70-215/8 70-215/9 70-215/10 70-225/7 70-225/8 70-225/9 70-225/10
112-3557EA 13.4 EUR
112-3557 112-3558 112-3559 112-3556 112-3561 112-3562 112-3563 112-3560
Snijbestendige handschoenen, HyFlex® 70-215 / 70-225
Handschoenen
These DuPont™ Kevlar® knitted gloves are designed to provide superior mechanical strength in light to medium duty handling and production applications.
  • Protection from sharp edges of metals, ceramics, glass and other materials
  • Reversible design allows glove to be worn on either hand, reducing replacement costs
  • Launderable up to 40 °C
Recommended for: Metalworking & assembly operations; chassis assembly, body trimming; seizing structural and body parts; inspection (picking, checking and marking); light duty welding applications
EN 388, EN 407 (X1XXXX), Cat. III
Certificaten: 70-215: EN 388:2016 (144XB), EN 407 (X1XXXX), Cat. III
70-225: EN 388:2016 (144XD), EN 407 (X1XXXX), EN 407 (4142XX), Cat. III