Handschoenen voor algemeen gebruik, Ultrane, 553
Leverancier: MAPA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34553087 34553088 34553089 34553080 34553086
112-0737EA 43.8 EUR
112-0737 112-0738 112-0739 112-0735 112-0736
Handschoenen voor algemeen gebruik, Ultrane, 553
Handschoenen
Dark grey polyamide knitted gloves with palm and fingers coated with black nitrile. Gloves feature curved fingers and contoured palm, elastic knitted wrist (natural rubber elastic wrist band) and smooth external surface. Guaranteed silicone free.
  • High abrasion resistance for enhanced durability
  • Nitrile coating offers superior resistance to oily or greasy components
  • Enhanced finger sensitivity with excellent precision of hand movement due to glove design and thickness
  • High comfort of seamless knitted liner, palm coated design for improved glove ventilation
These gloves are intended to protect the hand only. They do not protect the wrist or the forearm.
Certificaten: EN 388 (4121X), Cat. II