Ruimzichtbrillen, uvex ultrasonic
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9302.285 9302.600 9302.255 9302.245 9302.281 9302.286 9302.275
111-8009EA 37.8 EUR
111-8009 111-1911 111-8222 111-8008 111-8064 UVEX9302.286 111-1673
Ruimzichtbrillen, uvex ultrasonic
Ruimzichtbrillen
These full view goggles fit individual facial features well, due to their combination of hard and soft materials, ensuring a high level of comfort for the wearer.
  • Optimised internal volume and extensive side protection, even for spectacle wearers
  • Prescription insert possible
  • Wide panoramic field of view
  • Ventilated for a pleasant and irritation-free interior environment
Gewicht: 80 g.
* Model is voorzien van gereduceerde ventilatie boven en onder ** Model is voorzien van rubberen hoofdband.
** Model has rubber headband.
Certificaten: EN 166, EN 170; 1B.