Spiritusbrander, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
294020007 294030008
451-3111EA 0.9 EUR
451-3111 451-3112
Spiritusbrander, DURAN®
Verwarmers Alcohollampen
Soda-lime glass with ground glass joint cap. The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.
Inhoud: 100 ml.
Ø×H: 75×103 mm.
Bestelinformatie: Geleverd zonder vultuit en pit.