Biopsie cassettes in QuickLoad™ Sleeves, Micromesh™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M507-12SL M507-11SL M507-2SL M507-3SL M507-4SL M507-5SL M507-6SL M507-7SL M507-8SL M507-9SL M507-10SL
720-1224EA 321 EUR
720-1224 720-1578 720-1228 720-1229 720-1230 720-1231 720-1232 720-1582 720-1583 720-1584 720-1223
Biopsie cassettes in QuickLoad™ Sleeves, Micromesh™
Histologie-cassettes Cassettes voor biopsiebehandeling
Cassettes in plastic sleeves.
  • Anterior writing area is at a 45° angle to make the cassette more suitable for use with automated cassette printers
  • Available in non cytotoxic, non metallic colours
  • Perfect even for needle biopsies
  • Large labelling areas are located on three sides of the cassettes
Moulded from a special high density polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The lid can be opened and closed as often as necessary and always relocks securely without danger of specimen loss. Large square 27 mm compartment with 6 mm height.
Verpakking: Cassettes are packed in sleeves of 75. Each case contains 10 sleeves.