Monsternameborstels, Cytotak
Leverancier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MW146 MW147
720-0134EA 202 EUR
720-0134 720-0135
Monsternameborstels, Cytotak
Borstels
Nylon-bristle cytology sampling brushes for cervical smears.
  • Plastic shaft is non toxic and safe to use
  • Tapered spiral configuration assists access to narrow opening
  • Gentle rotation creates a brushing action against skin surface for enhanced recovery of cellular material
Shaft/head: Plastic/nylon
Verpakking: Available bulk or individually packed.
Opgelet: *The product(s) on this page are outside the scope of the EU Directive 2019/904