Korte aërometers
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3600PG070/20QP 3600PG136/20QP 3600PG142/20QP 3600PG148/20QP 3600PG154/20QP 3600PG160/20QP 3600PG166/20QP 3600PG172/20QP 3600PG178/20QP 3600PG076/20QP 3600PG082/20QP 360.088PMG 3600PG094/20QP 3600PG100/20QP 3600PG106/20QP 3600PG112/20QP 3600PG118/20QP 3600PG124/20QP 3600PG130/20QP
ALLA3600PG070/20QPEA 20.3 EUR
ALLA3600PG070/20QP 614-1654 614-1655 614-1656 ALLA3600PG154/20QP ALLA3600PG160/20QP ALLA3600PG166/20QP ALLA3600PG172/20QP ALLA3600PG178/20QP 614-0204 614-0205 614-0206 614-0207 614-0208 614-0209 614-0210 614-0211 614-0212 614-0213
Korte aërometers
Hydrometers
General purpose hydrometers calibrated at 20 °C.
  • Reading below the meniscus
Lengte 160 mm