Aërometers voor de petroleumindustrie
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3503LC10520BQP 350L060I649SPB 350L065I649SPB 350L070I649SPB 350L075I649SPB 350L080I649SPB 350L085I649SPB 350L090I649SPB 350L095I649SPB 350L100I649SPB
614-0505EA 86.5 EUR
614-0505 614-0504 614-0503 614-0502 614-0501 614-0500 614-0509 614-0508 614-0507 614-0506
Aërometers voor de petroleumindustrie
Hydrometers
These hydrometers for the petroleum industry have been designed with reading below the meniscus.
Certificaten: ISO 649, NF 35511, BS 718, DIN 12791.
Bestelinformatie: Traceable® three point calibration works certificate or official laboratory three point calibration certificate are available.
Order Now
Specificaties
Environmentally Preferable
Lengte 335 mm