Hydrometers, Brix
Leverancier: GERING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0669 0670 0671 0674S 0675S 0676S 0677S 0678WG 0679WG 0680WG 0681WG 0682SWG 0683SWG 0684SWG 0685SWG 0686SWG 0687 0688 0689 0690SWG 0691SWG 0692SWG
635-0314EA 42.4 EUR
635-0314 614-0070 614-0071 614-0072 614-0073 614-0074 614-0075 635-0315 635-0316 635-0317 614-0076 614-0077 614-0078 614-0079 614-0080 614-0081 635-0318 614-0082 635-0319 635-0320 614-0083 614-0084
Hydrometers, Brix
Hydrometers
Brix saccharimeters with a reference temperature of 20 °C.