Labels for Minimark printer, type B-595
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113204 113205 113182 113197 113210 113209 113208 113206 113202 113201 113198 113218 113193 113196 113194 113191 113185 113184 113200 113216 113217 115753 115751 113219 113215 113187 115760 115761 113212
BRDY113204EA 487.98 EUR
BRDY113204 BRDY113205 BRDY113182 BRDY113197 BRDY113210 BRDY113209 BRDY113208 BRDY113206 BRDY113202 BRDY113201 BRDY113198 BRDY113218 BRDY113193 BRDY113196 BRDY113194 BRDY113191 BRDY113185 BRDY113184 BRDY113200 BRDY113216 BRDY113217 BRDY115753 BRDY115751 BRDY113219 BRDY113215 811-0179 BRDY115760 BRDY115761 BRDY113212
Labels for Minimark printer, type B-595
Etiketten
These indoor/outdoor labeling tapes are made of vinyl film with permanent acrylic adhesive.
  • Sticks to highly textured surfaces
  • Adheres to difficult low surface energy items
  • 8 to 10 year outdoor durability