Halfgelaatsmakser, serie 7700
Leverancier: Honeywell Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N65770033 N65770032 N65770031
111-0289EA 48.9 EUR
111-0289 111-0290 111-0291
Halfgelaatsmakser, serie 7700
Ademhalingsmaskers Herbruikbare maskers Halfgelaatsmaskers
Soft, hypo-allergenic silicone half masks. Interchangeable cartridges and filters provide air-purifying protection for a variety of applications. Direct cartridge-to-face-piece connection eliminates gaskets and cartridge receptacles.
  • Soft silicone makes these respirators close fitting and comfortable
  • Fit snugly while the cradle suspension system alleviates pressure points
  • Can be worn with safety glasses and goggles
Certificaten: NIOSH approved for impact and penetration resistance when used with North Safety cartridges and/or filters.
Bestelinformatie: Cartridges and filters must be ordered separately