Schroeffilter
Leverancier: BARTELS RIEGER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
920600 920601 920603 920605 920850 920851 924400 924406 924700 924702 920650 924350
111-3072EA 14.9 EUR
111-3072 111-3073 111-3075 111-3076 111-3077 111-3078 111-3079 111-3081 111-3085 BART924702 BART920650 BART924350
Schroeffilter
Ademhalingsmaskers Herbruikbare maskers Maskerfilters
Standardised respiratory filter.
  • Suitable for masks with standard EN 148-1 round-thread connections
EN 141, 143 and 371.