Maatcilinders
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CT0010P CT0025P CT0050P CT0100P CT0250P CT0500P CT1000P CT2000P
612-0426EA 8.3 EUR
612-0426 612-0427 612-0428 612-0429 612-0430 612-0431 612-0432 612-0433
Maatcilinders
Cilinders Maatcilinders
PMP (TPX®), transparent, tall form, autoclavable, class B.
  • Hexagonal base with non-slip, anti-suction feature, spout, excellent chemical resistance, no meniscus to confuse readings
  • Recycling codes and batch code on each cylinder
  • Autoclavable - Does not affect accuracy
Note: The product is suitable for lab washing machines, however, a temperature of below 60°C should be used to prevent damage.
Certificaten: ISO 6706