Measuring cylinders, low form, class B, borosilicate glass, heavy duty, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3285-10 3285-25 3285-50 3285-100 3285-1L 3285-250 3285-500
612-0205EA 71.8 EUR
612-0205 612-0208 612-0210 612-0206 612-0207 612-0209 612-0211
Measuring cylinders, low form, class B, borosilicate glass, heavy duty, PYREX®
Cilinders Maatcilinders
Measuring cylinder, Borosilicate glass, low form, with spout.
  • Approximately half the height of equivalent standard cylinder for convenience and ease of use in confined spaces such as fume cupboards
  • Reinforced rims, thicker walls and robust hexagonal foot
  • Graduations in permanent blue enamel