Maatcilinders, met tuit, MBL
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CRC213 CRC202 CRC204 CRC206 CRC209 CRC211 CRC214 CRC216 CRC201
612-0423EA 67.9 EUR
612-0423 612-0418 612-0033 612-0034 612-0421 612-0422 612-0424 612-0425 612-4893
Maatcilinders, met tuit, MBL
Cilinders Maatcilinders
Borosilicate glass, tall form, class B.
  • Graduations in contrast blue enamel
  • Robust hexagonal foot
BS 604, ISO 4788, DIN 12680
Certificaten: BS 604, ISO 4788, DIN 12680
Order Now
Specificaties
Environmentally Preferable