Gesloten meetcilinder
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3215/06MP 3215/08P 3215/10P 3215/12P
BIBB3215/06MPEA 49.5 EUR
BIBB3215/06MP 612-0086 612-0088 612-0090
Gesloten meetcilinder
Cilinders Maatcilinders
These Class B cylinders comprise a PYREX® borosilicate glass cylinder with a polyethylene stopper and detachable polyethylene foot.
  • White enamel graduations
  • Polyethylene foot absorbs impact and reduces breakage
Certificaten: Complies with ISO 4788.
Bestelinformatie: Replacement parts available separately.