Maatcilinders
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3200/06MP 3200/18MP 3200/04MP 3200/08MP 3200/10MP 3200/12MP 3200/14MP 3200/16MP 3200/02MP
613-0918EA 25.4 EUR
613-0918 612-0064 612-0057 612-0059 612-0060 612-0061 612-0062 612-0063 BIBB3200/02MP
Maatcilinders
Cilinders Maatcilinders
Borosilicate glass, tall form, class B.
  • With spout
  • Graduations in high contrast white enamel
  • Hexagonal bases on cylinders above 5 ml for greater stability
Certificaten: BS 604, ISO 4788, DIN 12 680