Maatcilinders, lage vorm
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CPS0100P CPS0250P CPS0500P CPS1000P
215-0084EA 69.4 EUR
215-0084 215-0085 215-0086 215-0087
Maatcilinders, lage vorm
Cilinders Maatcilinders
PP, highly translucent, short form.
  • Unique self-draining base eliminates water collection during washing in lab washing machines, removing all spillage, re-wetting and contamination issues
  • Good chemical and temperature resistance
  • Anti suction base ensures no problems when working on wet surfaces
Space saving squat design for increased stability on the bench and ease of handling particularly in restricted spaces. Clear with blue over-printed graduations, which are always visible. Ideal for all laboratories, whether industrial, pharmaceutical or university.