Measuring cylinders, low form, class B, borosilicate glass
Leverancier: Bohemia Cristal (Kavalierglass)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612-3779EA 26.5 EUR
612-3779 612-3798 612-3800 612-3778 612-3799
Measuring cylinders, low form, class B, borosilicate glass
Cilinders Maatcilinders
Borosilicate glass, low form, class B.
  • Hexagonal base
  • White graduations
Certificaten: ISO 4788, DIN 12 680-1