Verzamel- en analyseplaten, Whatman UNIPLATE™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7701-5102 7701-5200 7701-5500 7701-1651 7701-3250
734-2558EA 445 EUR
734-2558 734-2559 734-2561 WHAT7701-1651 734-2583
Verzamel- en analyseplaten, Whatman UNIPLATE™
Microplaten HTS-microplaten
The UNIPLATE™ series includes microplates with a range of well profiles, well volumes and well densities, made from diverse polymer materials. Most UNIPLATE™ microplates conform to the proposed SBS microplate standard and fit most microplate readers and automated plate handling devices.
They are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with UNIFILTER® microplates, as well as homogeneous assay techniques utilised in HTS.