Celcultuurschalen, CELLSTAR®
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
628160_600 664160_360 627160_740 639160_120 627170_740
391-3210EA 155 EUR
391-3210 391-3236 391-3205 391-3225 392-0370
Celcultuurschalen, CELLSTAR®
Cultuurschalen Celcultuurschalen
PS, cell culture-treated, sterile. Available with a variety of different dimensions and growth areas. The 57 cm² and 143 cm² dishes have an extra high profile, with a height of 20 mm. All dishes are certified free of RNase, DNase and human DNA, are non pyrogenic and non cytotoxic.
  • Physical surface treatment for improved cell adhesion
  • Vents ensure efficient gas exchange
  • Easy stacking