Celcultuur multiwell platen, CELLSTAR®
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
662160_100 662102_100 657160_100 665180_100 677180_100 665102_100 677102_100 657185_100
391-3370EA 155 EUR
391-3370 391-3375 700-1425 392-0049 392-0062 392-0047 392-0061 392-0036
Celcultuur multiwell platen, CELLSTAR®
Microplaten Celcultuurplaten
PS, with lid, sterile. Cell culture multiwell plates are available either with a physically treated surface (for adherent cell cultures) or with a hydrophobic surface (for suspension cultures and hybridoma cells). All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • High clarity and low autofluorescence
  • Lid enables optimal gas exchange with the lowest possible evaporation
  • Single position lids prevent cross contamination
  • Alphanumeric well labelling
  • Compatible with automated systems