Deksels voor 96 well microplaten
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
656170_100 656101_100 656171_100 656161_100
736-6552EA 64.3 EUR
736-6552 736-6550 736-6553 738-0028
Deksels voor 96 well microplaten
Deksels Microplaatdeksels
These high profile lids, height 9 mm, are designed to protect against contamination and evaporation during sample storage and cell culture. Lids without condensation rings are mainly used with non tissue culture treated microplates. Lids with condensation rings are recommended for cell culture applications, since they inhibit cross contamination.