96-well microtestplaten, Falcon®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
353072 353075 353077 353227 353296 353910 353936 351172 351177 353916 353219 353376 353377
734-0023EA 192 EUR
734-0023 734-0025 734-0027 734-0057 734-0396 734-0080 736-2027 734-0954 734-0955 734-1376 734-2480 732-2194 732-2195
96-well microtestplaten, Falcon®
Microplaten Celcultuurplaten
Falcon® 96-well microplates are available in a variety of configurations and surface chemistries to meet a wide range of applications with fluorescence, luminescence, radiometric or colorimetric endpoints.
  • Flat bottom, with lid
  • Black wells with clear bottom
  • Working volume: 25 to 340 µl
  • Convenient, peel-open packaging
  • Available with untreated or tissue culture-treated surface chemistries
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Corning® Primaria™ tissue culture treatment additionally incorporates nitrogen-containing functional groups that have been shown to improve attachment and spreading of some cell types.
Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates.
Colour: Clear