Microplate covers, CapMats
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
381070_50 381061_50
736-0111EA 233 EUR
736-0111 736-0110
Microplate covers, CapMats
Afdichtingen Afdichtingen van microtiterplaten
Made from ethyl vinyl acetate (EVA) or silicone, CapMats offer an alternative method for sealing 96 and 384 well microplates. All CapMats are free of RNase, DNase, and human DNA, are non-pyrogenic, and are suitable for PCR applications.
  • 96-well CapMats with round naps are suitable for sealing 1 ml and 0,5 ml 96-well MASTERBLOCK®, as well as standard 96-well PP microplates
  • 96-well CapMats with round naps can also be used to seal 96-well F-, U-, and µClear® bottom PS microplates, but not V-bottom PS microplates
  • 96-well CapMats with square naps are suitable