PCR-platen met omranding
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T323-384SKN T323-96SKN
211-3394EA 563 EUR
211-3394 211-3375
PCR-platen met omranding
Microplaten PCR-platen
PP. These thin-walled PCR plates are suitable for robotic handling, and are ideal for use with automated pipetting systems.
  • Each well of the 96-well plate can hold a volume of 100 µl, while the 384-well plate has a capacity of 40 µl per well, suitable for reaction volumes of between 2 and 30 µl
  • Area for bar coding or labelling on each side and top
  • Alphanumeric grid for easy sample identification
  • Flat surface for better sealing
Verpakking: Packed in tamperproof, resealable bags of 10 plates.
Order Now
Specificaties
Environmentally Preferable