PCR-platen
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T323-24N T323-24B T323-24G T323-24R T323-24Y T323-48B T323-48G T323-48N T323-48R T323-48Y T323-96B T323-96G T323-96N T323-96R T323-96Y
211-3346EA 70.8 EUR
211-3346 211-3400 211-3399 211-3398 211-3397 211-3391 211-3390 211-3389 211-3388 211-3387 211-3386 211-3385 211-3379 211-3378 211-3372
PCR-platen
Microplaten PCR-platen
PP. Thin walled PCR plates compatible with 0,2 ml thermal cyclers. The insides of the tubes are smooth and have an inert surface on which enzymes and nucleic acids do not bind.
  • Alphanumeric grid for better identification
  • Flexible plate for better fitting of tubes in the thermal block
  • Larger plates are easily cut into sections of 24, 32 or 48 tubes, but, for more convenience, pre-cut plates are also available with formats of 48 tubes (6×8) and 24 tubes (3×8)
  • The bottom right corner of the plate is cut away to facilitate orientation
  • Certified free of RNase, DNase and DNA
Verpakking: Packed in tamperproof, resealable bags of 10 plates.