PCR-platen, 96- en 384-well
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
785201_60 785290_60 652201_40 652290_100 652270_100 652260_100
731-0039EA 471 EUR
731-0039 731-0040 731-0037 731-3016 GREI652270_100 GREI652260_100
PCR-platen, 96- en 384-well
Microplaten PCR-platen
PP, non-sterile, without lid. The use of the 96-well format allows the scale up of basic PCR work, while the 384-well format is ideal for high throughput screening projects, such as the sequencing of complex genomes.
  • Ultra thin PP for optimal heat transfer
  • Available in different designs, including half-skirted 96-well plates and full-skirted 384-well plates with one notch, suitable for ABI systems
  • Alphanumeric coding on 384-well plates, printed in colour
  • Free of DNase, RNase and human DNA, non-pyrogenic
Colour: Natural
Colour: Natural