Silicone aandrukmatten, Axygen®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CM-96-EXP CM-96-RD CM-FLAT
732-0672EA 60.6 EUR
732-0672 732-0671 732-0670
Silicone aandrukmatten, Axygen®
Afdichtingen Afdichtingen van microtiterplaten
Silicone sealing mats suitable for deep well plates and for thermal cyclers with screw down lids.
  • Prevents sample evaporation and reduces the risk of well-to-well contamination
  • Autoclavable and reusable
  • ImperaMat™ mats have higher chemical resistance
  • Temperature range: −80 to +121 °C
96-well expansion mats have a domed surface to ensure complete coverage of each well.