Objectglaasjes, Menzel Gläser
Leverancier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AA00000102E01 AB00000112E01 AA00000112E01 AB00000102E012
631-0701EA 4.2 EUR
631-0701 630-2012 630-1985 MENZAB00000102E012
Objectglaasjes, Menzel Gläser
Object- en dekglaasjes Objectglaasjes
These microscope slides are available plain or with one frosted end.
  • Vooraf was en polijsten
Conform to ISO 8037/I
Certificaten: ISO 8037/I