Flesjes op objectglaasjes, Nunc™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170920 177453
734-2107EA 682 EUR
734-2107 732-2609
Flesjes op objectglaasjes, Nunc™
Object- en dekglaasjes Objectglaasjes met cultuurkamers
Glass (Flaskette), PS (SlideFlask), sterile for cell culture on a microscope slide.
  • PS SlideFlask Nunclon™Δ certified
  • SlideFlask is ultrasonically welded to the slide, and individually leak tested
For karyotyping of cells, single cell autoradiography and single cell immuno-fluorescence.
W×D (Flaskette): 20×52 mm; W×D (SlideFlask): 18×50 mm