Moppen, Curtain Cleaner™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CCH-48 CCSC-2 CCSC-6 CCSC-6IR CCSC-612 CCSC-612IR
129-0501EA 1300 EUR
129-0501 129-0503 129-0504 129-0233 MICRCCSC-612 MICRCCSC-612IR
Moppen, Curtain Cleaner™
Moppen
A spring loaded, clamp mop that works to clean and sanitize vinyl strip curtains by clamping at the top and sliding down, effectively cleaning both sides at once. Ideal for critical environments, cleanroom support, and industrial applications.
  • Cover pairs fit over two, 305 mm bar clamps
  • Can be used with strong disinfectants
  • Launderable for re-use and autoclavable
Covers available in a choice of fabrics for specialised cleaning:
NovaPoly™ - smooth knit polyester provide excellent disinfectant dispersal.
PolySorb™ - textured polyester for removing residues and dispersing disinfectant.
NovaLite™ - latex bonded with rayon for greater friction resistance and absorbency.
The CurtainCleaner™ tool provides a simple, effective method to clean and sanitise cleanroom strip curtains and softwall enclosures, without the need for a ladder or stool. The durable stainless steel assembly includes an attached, 1200 mm, stainless steel handle with precise clamp hand control.
Bestelinformatie: Mop assembly and slipcovers must be ordered separately.