Quick disconnect handles, stainless steel
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SSU-3 SSA-3 SSU-1 SSU-7 SSU-5 SSU-6 SSU-4 SSU-2 SSU-8 SSU-9
129-0261EA 425 EUR
129-0261 129-0262 129-0551 129-0176 129-0737 129-0263 129-0062 129-0643 129-0644 129-0738
Quick disconnect handles, stainless steel
Moppen
Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.
  • Universal handles work with all Quick Disconnect tools and mop adapters
  • 304 electropolished stainless steel with polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available in a range of fixed or extendable lengths from 200 mm to 3 m
  • Quick Disconnect end attaches directly to QD mops, tools and adapters
  • SSA-3 adapter connects QD handles to plastic threaded mops, tools and adapters
  • Use the Adjustable Angle Adapters to reach over, under and around obstacles in the cleanroom
  • Good compatibility with disinfectants
  • Autoclavable
The Micronova Stainless Steel Universal Handles attach directly to the stainless steel Quick Disconnect (QD series) adapters and tools. Use the SSA-3 Quick Disconnect Adapter to attach to the plastic ACME threaded (QP series) adapters and tools. The Adjustable Angle Adapters can be used to create an adjustable angled shape between a handle and mop adapter to clean above and below fixtures, as well as, inside isolators and containment hoods.