Bulb pipettes, VOLAC®, (one mark), class AS
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R371/NC/G/5 R371/NC/B/5 R371/WAC/B/5 R371/WAC/C/5 R371/WAC/D/5 R371/WAC/G/5 R371/WAC/H/5 R371/WAC/J/5 R371/NC/H/5 R371/NC/J/5 R371/NC/K/5
612-4855EA 42.7 EUR
612-4855 POULR371/NC/B/5 POULR371/WAC/B/5 POULR371/WAC/C/5 POULR371/WAC/D/5 POULR371/WAC/G/5 POULR371/WAC/H/5 POULR371/WAC/J/5 POULR371/NC/H/5 POULR371/NC/J/5 POULR371/NC/K/5
Bulb pipettes, VOLAC®, (one mark), class AS
Pipetten Volpipetten
Glass.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in amber stain
  • Colour coded, individually numbered
BS 1583 - ISO 648
Certificaten: BS 1583 - ISO 648