Pipetten met graduatie, klasse AS, Schellbach
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1110129 1110133 1110141 1110153 1110154 1110160 1110165 1110170
612-1054EA 10.5 EUR
612-1054 612-1055 612-1056 612-1057 612-1058 612-1059 612-1060 612-1009
Pipetten met graduatie, klasse AS, Schellbach
Pipetten Gegradueerde pipetten
AR-Glas®.
  • Calibrated to deliver 'Ex', conformity certified with dated batch identification
  • Marks and inscriptions in blue stain, graduated to the tip, with Schellbach-stripe
  • From 5 ml with a cotton plug upper end
DIN 12 697
Length: 360 mm ±5 mm except for 25 ml pipettes: 450 mm ±5 mm.
Certificaten: DIN EN ISO 835. Conformity certified (DE-M marking).