Graduated pipettes, type 2, Class B, MBL
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRA800B PRA802B PRA804B PRA806B PRA808B
612-3191EA 20 EUR
612-3191 612-3190 612-3189 612-3188 612-3187
Graduated pipettes, type 2, Class B, MBL
Pipetten Gegradueerde pipetten
Soda lime glass.
  • Calibrated to deliver from any graduation line down to zero at the jet
  • Graduations in permanent amber stain
BS 700, ISO 835
Certificaten: BS 700, ISO 835